Learn More
Meso Scale Discovery VPLEX Plus Ab38 Peptide 4G8
SDP
Catalog No. 5000082286
Encompass
The Meso Scale Discovery V-PLEX Plus A 38 Peptide Kit is designed to quantitatively measure A 38 levels in human samples such as CSF serum and plasma This electrochemiluminescence (ECL) immunoassay utilizes MSD 4G8 antibody-coated plates and SULFO-TAG detection antibodies to provide high sensitivity a broad dynamic range and low matrix interference The kit includes a plate detection antibody calibrator and assay diluent optimized for A 38 detection It allows rapid analysis of A 38 levels an important biomarker associated with Alzheimer s disease progression With minimal hands-on time this kit enables efficient and accurate A 38 quantification
Catalog No. 50-000-82286 Supplier Meso Scale Discovery Supplier No. K150SHG1
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX Plus A 38 Peptide Kit is designed to quantitatively measure A 38 levels in human samples such as CSF serum and plasma This electrochemiluminescence (ECL) immunoassay utilizes MSD 4G8 antibody-coated plates and SULFO-TAG detection antibodies to provide high sensitivity a broad dynamic range and low matrix interference The kit includes a plate detection antibody calibrator and assay diluent optimized for A 38 detection It allows rapid analysis of A 38 levels an important biomarker associated with Alzheimer s disease progression With minimal hands-on time this kit enables efficient and accurate A 38 quantification