Learn More
Meso Scale Discovery VPLEX Plus Ab38 Peptide 4G8
SDP
Catalog No. 5000082287
Encompass
The Meso Scale Discovery V-PLEX Plus A 38 Peptide Kit is designed to quantitatively measure A 38 levels in human samples such as CSF serum and plasma This electrochemiluminescence (ECL) immunoassay utilizes MSD 4G8 antibody-coated plates and SULFO-TAG detection antibodies to provide high sensitivity broad dynamic range and low background The kit includes a plate detection antibody calibrator diluent and wash buffer for 96 tests It can accurately quantify A 38 levels between 12 2-2000 pg/mL with high specificity and no cross-reactivity to A 40 or A 42 This research-use-only kit enables understanding of A peptide profiles related to Alzheimer s disease pathology
Catalog No. 50-000-82287 Supplier Meso Scale Discovery Supplier No. K150SHG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus A 38 Peptide Kit is designed to quantitatively measure A 38 levels in human samples such as CSF serum and plasma This electrochemiluminescence (ECL) immunoassay utilizes MSD 4G8 antibody-coated plates and SULFO-TAG detection antibodies to provide high sensitivity broad dynamic range and low background The kit includes a plate detection antibody calibrator diluent and wash buffer for 96 tests It can accurately quantify A 38 levels between 12 2-2000 pg/mL with high specificity and no cross-reactivity to A 40 or A 42 This research-use-only kit enables understanding of A peptide profiles related to Alzheimer s disease pathology