Learn More
Meso Scale Discovery VPLEX Plus Ab38 Peptide 6E10
SDP
Catalog No. 5000081602
Encompass
The Meso Scale Discovery V-PLEX A Peptide Panel 1 (6E10) Kit is a multiplex immunoassay that measures A 38 A 40 and A 42 in human samples such as CSF serum and plasma It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range low sample volume requirements and rapid results The assay requires 50 L of sample and takes 3 5 hours to complete It includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels and calibrators and controls The kit allows simultaneous measurement of A peptides which aids in understanding amyloid processing associated with Alzheimer s disease
Catalog No. 50-000-81602 Supplier Meso Scale Discovery Supplier No. K150SIG2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX A Peptide Panel 1 (6E10) Kit is a multiplex immunoassay that measures A 38 A 40 and A 42 in human samples such as CSF serum and plasma It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range low sample volume requirements and rapid results The assay requires 50 L of sample and takes 3 5 hours to complete It includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels and calibrators and controls The kit allows simultaneous measurement of A peptides which aids in understanding amyloid processing associated with Alzheimer s disease