Learn More
Meso Scale Discovery VPLEX Plus Ab40 Peptide 4G8
SDP
Catalog No. 5000078313
Encompass
The Meso Scale Discovery V-PLEX Plus Abeta40 Peptide Kit measures Abeta40 in human samples like serum plasma CSF and cell culture supernatants This ECL immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and low sample volume It includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are analyzed on MSD s platform providing quantitation of Abeta40 concentration allowing rapid reproducible results from small samples
Catalog No. 50-000-78313 Supplier Meso Scale Discovery Supplier No. K150SJG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Abeta40 Peptide Kit measures Abeta40 in human samples like serum plasma CSF and cell culture supernatants This ECL immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and low sample volume It includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are analyzed on MSD s platform providing quantitation of Abeta40 concentration allowing rapid reproducible results from small samples