Learn More
Meso Scale Discovery VPLEX Plus Ab40 Peptide 4G8
SDP
Catalog No. 5000078322
Encompass
The V-PLEX Plus A 40 Peptide Kit (K150SJG) from Meso Scale Discovery is an immunoassay that quantitatively measures A 40 levels in human CSF and plasma samples It utilizes electrochemiluminescence detection technology and provides high sensitivity a broad dynamic range and requires low sample volumes The kit includes a 96-well plate coated with a monoclonal antibody specific for the N-terminus of A 40 a detection antibody targeting the C-terminus SULFO-TAG labeled streptavidin wash buffers blocking solution calibrators and positive controls It allows rapid quantification of A 40 levels with a testing time of just 3 5 hours
Catalog No. 50-000-78322 Supplier Meso Scale Discovery Supplier No. K150SJG2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus A 40 Peptide Kit (K150SJG) from Meso Scale Discovery is an immunoassay that quantitatively measures A 40 levels in human CSF and plasma samples It utilizes electrochemiluminescence detection technology and provides high sensitivity a broad dynamic range and requires low sample volumes The kit includes a 96-well plate coated with a monoclonal antibody specific for the N-terminus of A 40 a detection antibody targeting the C-terminus SULFO-TAG labeled streptavidin wash buffers blocking solution calibrators and positive controls It allows rapid quantification of A 40 levels with a testing time of just 3 5 hours