Learn More
Meso Scale Discovery VPLEX Plus Ab40 Peptide 6E10
SDP
Catalog No. 5000078324
Encompass
The V-PLEX Plus A 40 Peptide Kit from Meso Scale Discovery is used to quantitatively measure amyloid beta 40 (A 40) in various sample types such as serum plasma and cell culture supernatants This electrochemiluminescence (ECL) immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results comparable to ELISA The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay With a broad dynamic range and high sensitivity this kit is well-suited for studying A 40 levels in Alzheimer s disease research
Catalog No. 50-000-78324 Supplier Meso Scale Discovery Supplier No. K150SKG2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus A 40 Peptide Kit from Meso Scale Discovery is used to quantitatively measure amyloid beta 40 (A 40) in various sample types such as serum plasma and cell culture supernatants This electrochemiluminescence (ECL) immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results comparable to ELISA The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay With a broad dynamic range and high sensitivity this kit is well-suited for studying A 40 levels in Alzheimer s disease research