Learn More
Meso Scale Discovery VPLEX Plus Ab42 Peptide 4G8
SDP
Catalog No. 5000080578
Encompass
The K150SLG-1 is a 150mm diameter silicon wafer produced by Mesoscale Technologies It is a prime grade single side polished wafer with a thickness of 725 m /- 25 m The wafer has a resistivity of 1-10 Ohm-cm orientation of 100 and is single side polished to a surface roughness of 1nm Ra As a prime grade wafer it has minimal defects with 5 particles 0 2 m per cm 2 The K150SLG-1 is well-suited for microfabrication processes requiring a high quality single crystal silicon substrate
Catalog No. 50-000-80578 Supplier Meso Scale Discovery Supplier No. K150SLG1
The K150SLG-1 is a 150mm diameter silicon wafer produced by Mesoscale Technologies It is a prime grade single side polished wafer with a thickness of 725 m /- 25 m The wafer has a resistivity of 1-10 Ohm-cm orientation of 100 and is single side polished to a surface roughness of 1nm Ra As a prime grade wafer it has minimal defects with 5 particles 0 2 m per cm 2 The K150SLG-1 is well-suited for microfabrication processes requiring a high quality single crystal silicon substrate