Learn More
Meso Scale Discovery VPLEX Plus Chemokine PNL NHP
SDP
Catalog No. 5000081824
Encompass
Meso Scale Discovery offers the V-PLEX NHP Cytokine Panel 30-Plex Kit for the quantitative measurement of 30 non-human primate cytokines chemokines and growth factors This multiplex assay allows researchers to measure biomarkers from small sample volumes with high sensitivity and broad dynamic range The kit includes a 30-plex primate panel calibrators controls detection antibodies read buffer sample diluent and plates It utilizes electrochemiluminescence technology on Meso Scale Discovery s instrumentation platforms for high performance immunoassays This product enables efficient analysis of NHP biomarkers relevant to modeling human diseases
Catalog No. 50-000-81824 Supplier Meso Scale Discovery Supplier No. K15055G4
Add to Cart
Add to Cart
Request a Quote
Meso Scale Discovery offers the V-PLEX NHP Cytokine Panel 30-Plex Kit for the quantitative measurement of 30 non-human primate cytokines chemokines and growth factors This multiplex assay allows researchers to measure biomarkers from small sample volumes with high sensitivity and broad dynamic range The kit includes a 30-plex primate panel calibrators controls detection antibodies read buffer sample diluent and plates It utilizes electrochemiluminescence technology on Meso Scale Discovery s instrumentation platforms for high performance immunoassays This product enables efficient analysis of NHP biomarkers relevant to modeling human diseases