Learn More
Meso Scale Discovery V-PLEX Plus Cytokine Panel 1 Mouse Kit, 96-well plate, Multiplex Immunoassay, Electrochemiluminescence (ECL), 9 analytes, Serum, Plasma, Cell Culture Supernatant, Urine
SDP
Supplier:  Meso Scale Discovery K15245G1
Encompass
The MESO SCALE DISCOVERY V-PLEX Plus Cytokine Panel 1 Mouse Kit is a highly sensitive multiplex immunoassay designed for the quantitative measurement of nine key biomarkers in mouse samples. This kit is ideal for research in inflammatory responses and immune system regulation, utilizing a proprietary electrochemiluminescence (ECL) platform for reliable and reproducible data collection.
  • Multiplexing capability enables simultaneous quantification of nine distinct cytokine biomarkers.
  • High sensitivity with detection limits in the sub- to low pg/mL range.
  • Rigorous validation ensures lot-to-lot consistency for reliable results.
  • Compatible with various mouse sample types, including serum and plasma.
  • Comprehensive kit components streamline assay setup.
Catalog No. NC3356386