Learn More
Meso Scale Discovery VPLEX Plus Cytokine PNL mouse
SDP
Catalog No. 5000082355
Encompass
The V-PLEX Plus Cytokine Panel 1 (Mouse) Kit from Meso Scale Discovery is a multiplex immunoassay that allows simultaneous measurement of 10 mouse cytokines and chemokines (GM-CSF IFN- IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 and TNF- ) from cell culture supernatants serum plasma and other biological fluids The kit utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The 10-plex panel allows efficient profiling of cytokines and chemokines involved in innate and adaptive immune responses
Catalog No. 50-000-82355 Supplier Meso Scale Discovery Supplier No. K15245G4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Cytokine Panel 1 (Mouse) Kit from Meso Scale Discovery is a multiplex immunoassay that allows simultaneous measurement of 10 mouse cytokines and chemokines (GM-CSF IFN- IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 and TNF- ) from cell culture supernatants serum plasma and other biological fluids The kit utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The 10-plex panel allows efficient profiling of cytokines and chemokines involved in innate and adaptive immune responses