Learn More
Meso Scale Discovery VPLEX Plus Cytokine PNL NHP
SDP
Catalog No. 5000078811
Encompass
Meso Scale Discovery offers a non-human primate interleukin 7 (IL-7) assay kit to quantitatively measure IL-7 levels in serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology for high sensitivity and wide dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent and vials of calibrator to generate a standard curve The assay takes 3 5 hours for completion and has a lower limit of detection of 0 3 pg/mL IL-7
Catalog No. 50-000-78811 Supplier Meso Scale Discovery Supplier No. K15057G2
Add to Cart
Add to Cart
Request a Quote
Meso Scale Discovery offers a non-human primate interleukin 7 (IL-7) assay kit to quantitatively measure IL-7 levels in serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology for high sensitivity and wide dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent and vials of calibrator to generate a standard curve The assay takes 3 5 hours for completion and has a lower limit of detection of 0 3 pg/mL IL-7