Learn More
Meso Scale Discovery VPLEX Plus Cytokine PNL NHP
SDP
Catalog No. 5000082279
Encompass
The Meso Scale Discovery V-PLEX Plus Cytokine Panel 1 NHP Kit is a multiplex immunoassay measuring 10 non-human primate cytokines and chemokines (IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IFN- TNF- ) from various biological fluids It uses electrochemiluminescence detection for high sensitivity and broad range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals for one plate enabling efficient NHP cytokine analysis
Catalog No. 50-000-82279 Supplier Meso Scale Discovery Supplier No. K15057G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Cytokine Panel 1 NHP Kit is a multiplex immunoassay measuring 10 non-human primate cytokines and chemokines (IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IFN- TNF- ) from various biological fluids It uses electrochemiluminescence detection for high sensitivity and broad range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals for one plate enabling efficient NHP cytokine analysis