Learn More
Meso Scale Discovery VPLEX Plus GIP active
SDP
Catalog No. 5000078878
Encompass
The Meso Scale Discovery V-PLEX Plus GIP Active Kit is a 96-well plate assay for measuring active GLP-1 levels in serum plasma and cell culture supernatants This electrochemiluminescence immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantification of active GLP-1 down to 0 15 pg/mL The kit includes a 96-well plate with a GLP-1 capture antibody detection antibody with MSD SULFO-TAG calibrators and an optimized assay diluent It enables high-throughput analysis of active GLP-1 levels with a 3 5-hour protocol
Catalog No. 50-000-78878 Supplier Meso Scale Discovery Supplier No. K150T5G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus GIP Active Kit is a 96-well plate assay for measuring active GLP-1 levels in serum plasma and cell culture supernatants This electrochemiluminescence immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantification of active GLP-1 down to 0 15 pg/mL The kit includes a 96-well plate with a GLP-1 capture antibody detection antibody with MSD SULFO-TAG calibrators and an optimized assay diluent It enables high-throughput analysis of active GLP-1 levels with a 3 5-hour protocol