Learn More
Meso Scale Discovery VPLEX Plus GLP1 Active
SDP
Catalog No. 5000081670
Encompass
The Meso Scale Discovery V-PLEX Plus GLP-1 Active Kit quantitatively measures active GLP-1 in serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 96-well plate with a GLP-1 capture antibody a detection antibody with an MSD SULFO-TAG and calibrators for a standard curve It detects 1 5-1000 pg/mL and shows no cross-reactivity with related analytes The assay runs on MSD s SECTOR Imager 6000
Catalog No. 50-000-81670 Supplier Meso Scale Discovery Supplier No. K1503OG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus GLP-1 Active Kit quantitatively measures active GLP-1 in serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 96-well plate with a GLP-1 capture antibody a detection antibody with an MSD SULFO-TAG and calibrators for a standard curve It detects 1 5-1000 pg/mL and shows no cross-reactivity with related analytes The assay runs on MSD s SECTOR Imager 6000