Learn More
Meso Scale Discovery VPLEX Plus GLP1 Total
SDP
Catalog No. 5000078250
Encompass
The Meso Scale Discovery V-PLEX Plus GLP-1 Total Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total glucagon-like peptide-1 (GLP-1) in human serum plasma and culture media samples The kit contains a 96-well plate coated with a GLP-1 specific capture antibody a ruthenium-labeled detection antibody calibrators controls assay diluent read buffer wash buffer and plate seals The assay has a range from 0 61 pg/mL to 5000 pg/mL and takes approximately 3 5 hours to complete This product allows for the sensitive and accurate quantification of total GLP-1
Catalog No. 50-000-78250 Supplier Meso Scale Discovery Supplier No. K1503PG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus GLP-1 Total Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total glucagon-like peptide-1 (GLP-1) in human serum plasma and culture media samples The kit contains a 96-well plate coated with a GLP-1 specific capture antibody a ruthenium-labeled detection antibody calibrators controls assay diluent read buffer wash buffer and plate seals The assay has a range from 0 61 pg/mL to 5000 pg/mL and takes approximately 3 5 hours to complete This product allows for the sensitive and accurate quantification of total GLP-1