Learn More
Meso Scale Discovery VPLEX Plus Glucagon
SDP
Catalog No. 5000081633
Encompass
The Meso Scale Discovery V-PLEX Plus Glucagon Kit is a highly sensitive immunoassay that quantitatively measures glucagon levels in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide superior sensitivity a wide dynamic range and requires just 50 L of sample The V-PLEX Plus Glucagon Kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and diluent This assay kit can accurately quantify glucagon levels between 0 17-1500 pg/mL
Catalog No. 50-000-81633 Supplier Meso Scale Discovery Supplier No. K150U5G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Glucagon Kit is a highly sensitive immunoassay that quantitatively measures glucagon levels in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide superior sensitivity a wide dynamic range and requires just 50 L of sample The V-PLEX Plus Glucagon Kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and diluent This assay kit can accurately quantify glucagon levels between 0 17-1500 pg/mL