Learn More
Meso Scale Discovery VPLEX Plus HUM CPeptide
SDP
Catalog No. 5000081740
Encompass
The Meso Scale Discovery V-PLEX Human C-Peptide Kit is an electrochemiluminescent immunoassay for the quantitative measurement of C-peptide in human serum plasma and culture media samples The assay utilizes MSD s proprietary V-PLEX technology is designed for use on MSD instrumentation including QuickPlex SQ 120 and has a detection range of 50-25 000 pg/mL The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay has high sensitivity wide dynamic range and requires low sample volume
Catalog No. 50-000-81740 Supplier Meso Scale Discovery Supplier No. K151X5G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human C-Peptide Kit is an electrochemiluminescent immunoassay for the quantitative measurement of C-peptide in human serum plasma and culture media samples The assay utilizes MSD s proprietary V-PLEX technology is designed for use on MSD instrumentation including QuickPlex SQ 120 and has a detection range of 50-25 000 pg/mL The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay has high sensitivity wide dynamic range and requires low sample volume