Learn More
Meso Scale Discovery VPLEX Plus HUM CPeptide Kit
SDP
Catalog No. 5000079752
Encompass
The Meso Scale Discovery V-PLEX Human C-Peptide Kit is an electrochemiluminescent immunoassay for the quantitative measurement of C-peptide in human serum plasma and culture media samples The assay utilizes MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of 78-20 000 pg/mL and shows minimal cross-reactivity The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and reagents required to perform the assay Sample incubation time is 2 hours The kit can aid in monitoring endogenous insulin production and has applications in diabetes research
Catalog No. 50-000-79752 Supplier Meso Scale Discovery Supplier No. K151X5G2
The Meso Scale Discovery V-PLEX Human C-Peptide Kit is an electrochemiluminescent immunoassay for the quantitative measurement of C-peptide in human serum plasma and culture media samples The assay utilizes MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of 78-20 000 pg/mL and shows minimal cross-reactivity The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and reagents required to perform the assay Sample incubation time is 2 hours The kit can aid in monitoring endogenous insulin production and has applications in diabetes research