Learn More
Meso Scale Discovery VPLEX Plus HUM CPeptide Kit
SDP
Catalog No. 5000081922
Encompass
The Meso Scale Discovery V-PLEX C-Peptide Kit is a singleplex immunoassay for quantifying human C-Peptide in serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of C-Peptide levels as low as 0 008 ng/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents It is ideal for monitoring insulin production and diabetes research offering high sensitivity wide dynamic range and minimal matrix interference
Catalog No. 50-000-81922 Supplier Meso Scale Discovery Supplier No. K151X5G1
The Meso Scale Discovery V-PLEX C-Peptide Kit is a singleplex immunoassay for quantifying human C-Peptide in serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of C-Peptide levels as low as 0 008 ng/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents It is ideal for monitoring insulin production and diabetes research offering high sensitivity wide dynamic range and minimal matrix interference