Learn More
Meso Scale Discovery VPLEX Plus HUM Cytokine 29Plex
SDP
Supplier:  Meso Scale Discovery K15054G2
Encompass
The V-PLEX Plus Human Cytokine 30-plex Kit from Meso Scale Discovery is a multiplex immunoassay that allows simultaneous measurement of 30 human cytokines chemokines and growth factors in a single small sample volume The kit contains a 96-well plate coated with capture antibodies specific for each analyte detection antibodies conjugated to an electrochemiluminescent label and calibrator solutions for generating standard curves The assay has high sensitivity broad dynamic range and requires only 25 L of sample per well This kit enables efficient profiling of cytokines and chemokines related to inflammation immune response and growth regulation
Catalog No. 50-000-78251
Only null left
Add to Cart
Request a Quote