Learn More
Meso Scale Discovery VPLEX Plus HUM Cytokine 29Plex
SDP
Catalog No. 5000078277
Encompass
The V-PLEX Plus Human Cytokine 30-plex Kit from Meso Scale Discovery is a multiplex immunoassay that allows simultaneous measurement of 30 human cytokines chemokines and growth factors in a single small sample volume The kit contains a 96-well plate coated with capture antibodies specific for each analyte detection antibodies conjugated to an electrochemiluminescent label and calibrator solutions for generating standard curves The assay has a rapid procedure of just 3 5 hours with no wash steps and provides limits of detection in the sub-pg/mL range This kit enables efficient profiling of cytokines and chemokines related to inflammation immune response and growth regulation
Catalog No. 50-000-78277 Supplier Meso Scale Discovery Supplier No. K15054G4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human Cytokine 30-plex Kit from Meso Scale Discovery is a multiplex immunoassay that allows simultaneous measurement of 30 human cytokines chemokines and growth factors in a single small sample volume The kit contains a 96-well plate coated with capture antibodies specific for each analyte detection antibodies conjugated to an electrochemiluminescent label and calibrator solutions for generating standard curves The assay has a rapid procedure of just 3 5 hours with no wash steps and provides limits of detection in the sub-pg/mL range This kit enables efficient profiling of cytokines and chemokines related to inflammation immune response and growth regulation