Learn More
Meso Scale Discovery VPLEX Plus HUM Cytokine 29Plex
SDP
Catalog No. 5000081593
Encompass
The V-PLEX Plus Human Cytokine 30-plex Kit from Meso Scale Discovery is a multiplex immunoassay for measuring 30 human cytokines chemokines and growth factors in a small sample It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrator solutions The assay is sensitive has a broad range and uses only 25 L per well Results are ready in 3 5 hours with an MSD instrument This kit efficiently profiles cytokines and chemokines related to inflammation immune response and growth regulation
Catalog No. 50-000-81593 Supplier Meso Scale Discovery Supplier No. K15054G1
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human Cytokine 30-plex Kit from Meso Scale Discovery is a multiplex immunoassay for measuring 30 human cytokines chemokines and growth factors in a small sample It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrator solutions The assay is sensitive has a broad range and uses only 25 L per well Results are ready in 3 5 hours with an MSD instrument This kit efficiently profiles cytokines and chemokines related to inflammation immune response and growth regulation