Learn More
Meso Scale Discovery VPLEX Plus HUM Eotaxin3 Kit
SDP
Catalog No. 5000079151
Encompass
The Meso Scale Discovery V-PLEX Human Eotaxin-3 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human Eotaxin-3 in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and vials of calibrator to generate a standard curve The assay has a detection range of 0 06-2500 pg/mL and takes 3 5 hours to complete The kit includes sufficient reagents for one 96-well plate and can assay 40 samples in duplicate
Catalog No. 50-000-79151 Supplier Meso Scale Discovery Supplier No. K151NUG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human Eotaxin-3 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human Eotaxin-3 in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and vials of calibrator to generate a standard curve The assay has a detection range of 0 06-2500 pg/mL and takes 3 5 hours to complete The kit includes sufficient reagents for one 96-well plate and can assay 40 samples in duplicate