Learn More
Meso Scale Discovery VPLEX Plus HUM Eotaxin3 Kit
SDP
Catalog No. 5000079603
Encompass
The Meso Scale Discovery V-PLEX Human Eotaxin-3 Kit is a highly sensitive immunoassay that quantitatively measures human Eotaxin-3 levels in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide superior sensitivity a wide dynamic range and requires just 50-100 L of sample The V-PLEX Eotaxin-3 Kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and assay diluents valid for serum EDTA plasma heparin plasma and cell culture supernatant samples
Catalog No. 50-000-79603 Supplier Meso Scale Discovery Supplier No. K151NUG2
The Meso Scale Discovery V-PLEX Human Eotaxin-3 Kit is a highly sensitive immunoassay that quantitatively measures human Eotaxin-3 levels in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide superior sensitivity a wide dynamic range and requires just 50-100 L of sample The V-PLEX Eotaxin-3 Kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and assay diluents valid for serum EDTA plasma heparin plasma and cell culture supernatant samples