Learn More
Meso Scale Discovery VPLEX Plus HUM Eotaxin3 Kit
SDP
Catalog No. 5000082474
Encompass
The Meso Scale Discovery V-PLEX Plus Human Eotaxin-3 Kit is a highly sensitive immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of human Eotaxin-3 in serum plasma and cell culture supernatants The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators and diluent The assay has a detection range of 0 03-2500 pg/mL and exhibits minimal cross-reactivity to related analytes This kit allows for the accurate quantification of Eotaxin-3 levels from various biological samples
Catalog No. 50-000-82474 Supplier Meso Scale Discovery Supplier No. K151NUG1
The Meso Scale Discovery V-PLEX Plus Human Eotaxin-3 Kit is a highly sensitive immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of human Eotaxin-3 in serum plasma and cell culture supernatants The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators and diluent The assay has a detection range of 0 03-2500 pg/mL and exhibits minimal cross-reactivity to related analytes This kit allows for the accurate quantification of Eotaxin-3 levels from various biological samples