Learn More
Meso Scale Discovery VPLEX Plus HUM FGF basic Kit
SDP
Catalog No. 5000079172
Encompass
The V-PLEX Human FGF Basic Kit is a multiplex immunoassay for measuring FGF basic in various biological fluids It uses ECL technology on the MSD platform for high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibodies detection antibody conjugates calibrators and controls It has a sensitivity of 1 5 pg/mL and an upper limit of 10 000 pg/mL requiring 50 L of sample per well The kit provides all necessary reagents to quantify FGF basic levels in human samples
Catalog No. 50-000-79172 Supplier Meso Scale Discovery Supplier No. K151MDG2
The V-PLEX Human FGF Basic Kit is a multiplex immunoassay for measuring FGF basic in various biological fluids It uses ECL technology on the MSD platform for high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibodies detection antibody conjugates calibrators and controls It has a sensitivity of 1 5 pg/mL and an upper limit of 10 000 pg/mL requiring 50 L of sample per well The kit provides all necessary reagents to quantify FGF basic levels in human samples