Learn More
Meso Scale Discovery VPLEX Plus HUM FGF basic Kit
SDP
Catalog No. 5000082235
Encompass
The Meso Scale Discovery K151MDG-1 is a human IL-1 beta assay kit that uses electrochemiluminescent detection technology for ultra-sensitive and reproducible quantification of IL-1 beta in serum plasma cell culture supernatants and other biological samples The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and assay diluent The assay has a detection range of 3 2-10 000 pg/mL and takes approximately 3 5 hours to complete It provides a sensitive and reliable way to measure IL-1 beta levels for life science research applications
Catalog No. 50-000-82235 Supplier Meso Scale Discovery Supplier No. K151MDG1
The Meso Scale Discovery K151MDG-1 is a human IL-1 beta assay kit that uses electrochemiluminescent detection technology for ultra-sensitive and reproducible quantification of IL-1 beta in serum plasma cell culture supernatants and other biological samples The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and assay diluent The assay has a detection range of 3 2-10 000 pg/mL and takes approximately 3 5 hours to complete It provides a sensitive and reliable way to measure IL-1 beta levels for life science research applications