Learn More
Meso Scale Discovery VPLEX Plus HUM FGF basic Kit
SDP
Catalog No. 5000082326
Encompass
The Meso Scale Discovery FGF basic Kit is a 4-plex immunoassay for measuring FGF basic (FGF2) in various biological samples It uses MSD s electrochemiluminescent detection for high sensitivity and a wide dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents Detection limits range from 1 5 to 10 000 pg/mL and the assay takes about 3 5 hours This product allows sensitive reproducible quantitation of FGF basic for life science research
Catalog No. 50-000-82326 Supplier Meso Scale Discovery Supplier No. K151MDG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery FGF basic Kit is a 4-plex immunoassay for measuring FGF basic (FGF2) in various biological samples It uses MSD s electrochemiluminescent detection for high sensitivity and a wide dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents Detection limits range from 1 5 to 10 000 pg/mL and the assay takes about 3 5 hours This product allows sensitive reproducible quantitation of FGF basic for life science research