Learn More
Meso Scale Discovery VPLEX Plus HUM IL12/IL23p40Pl
SDP
Catalog No. 5000078762
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-12 (p70) and IL-23 (p40) Kit is a multiplex immunoassay that allows simultaneous quantification of IL-12 (p70) and IL-23 (p40) in human serum plasma and culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators controls 10X diluent read buffer wash buffer and plate seals The assay takes approximately 3 hours to complete after a 2 hour incubation
Catalog No. 50-000-78762 Supplier Meso Scale Discovery Supplier No. K151RJG1
The Meso Scale Discovery V-PLEX Plus Human IL-12 (p70) and IL-23 (p40) Kit is a multiplex immunoassay that allows simultaneous quantification of IL-12 (p70) and IL-23 (p40) in human serum plasma and culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators controls 10X diluent read buffer wash buffer and plate seals The assay takes approximately 3 hours to complete after a 2 hour incubation