Learn More
Meso Scale Discovery VPLEX Plus HUM IL12 p70 Kit
SDP
Catalog No. 5000079879
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-12p70 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 12 (IL-12) p70 in cell culture supernatants serum and plasma The kit contains a 96-well plate coated with a capture antibody specific for human IL-12p70 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 6 pg/mL and a 7-log dynamic range from 0 6-10 000 pg/mL Sample incubation time is 2 hours
Catalog No. 50-000-79879 Supplier Meso Scale Discovery Supplier No. K151QVG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human IL-12p70 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 12 (IL-12) p70 in cell culture supernatants serum and plasma The kit contains a 96-well plate coated with a capture antibody specific for human IL-12p70 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 6 pg/mL and a 7-log dynamic range from 0 6-10 000 pg/mL Sample incubation time is 2 hours