Learn More
Meso Scale Discovery VPLEX Plus HUM IL12 p70 Kit
SDP
Catalog No. 5000081266
Encompass
The V-PLEX Plus Human IL-12p70 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 12 (IL-12) in serum plasma and culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody specific for human IL-12p70 a vial of SULFO-TAG labeled anti-human IL-12p40 detection antibody assay diluent read buffer wash buffer and a vial of lyophilized recombinant human IL-12p70 standard The sensitivity of the assay is 1 4 pg/mL with an assay range from 2 4-1500 pg/mL This product allows for the accurate quantitation of IL-12p70 levels in human samples
Catalog No. 50-000-81266 Supplier Meso Scale Discovery Supplier No. K151QVG2
Add to Cart
Add to Cart
The V-PLEX Plus Human IL-12p70 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 12 (IL-12) in serum plasma and culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody specific for human IL-12p70 a vial of SULFO-TAG labeled anti-human IL-12p40 detection antibody assay diluent read buffer wash buffer and a vial of lyophilized recombinant human IL-12p70 standard The sensitivity of the assay is 1 4 pg/mL with an assay range from 2 4-1500 pg/mL This product allows for the accurate quantitation of IL-12p70 levels in human samples