Learn More
Meso Scale Discovery VPLEX Plus HUM IL17A/F
SDP
Catalog No. 5000082398
Encompass
The Meso Scale Discovery Phospho (Ser473)/Total Akt Whole Cell Lysate Kit is designed to measure endogenous levels of Akt and phosphorylated Akt (Ser473) in whole cell lysates The kit includes a 96-well plate coated with capture antibodies against total Akt and phospho-Akt (Ser473) It also includes detection antibodies and buffers needed to perform the assay Results are obtained in as little as 3 5 hours using an electrochemiluminescence plate reader This kit allows for the sensitive reproducible quantification of Akt phosphorylation and activation state in cells It is suitable for studying Akt signaling and screening compounds that modulate Akt phosphorylation
Catalog No. 50-000-82398 Supplier Meso Scale Discovery Supplier No. K151WNG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Phospho (Ser473)/Total Akt Whole Cell Lysate Kit is designed to measure endogenous levels of Akt and phosphorylated Akt (Ser473) in whole cell lysates The kit includes a 96-well plate coated with capture antibodies against total Akt and phospho-Akt (Ser473) It also includes detection antibodies and buffers needed to perform the assay Results are obtained in as little as 3 5 hours using an electrochemiluminescence plate reader This kit allows for the sensitive reproducible quantification of Akt phosphorylation and activation state in cells It is suitable for studying Akt signaling and screening compounds that modulate Akt phosphorylation