Learn More
Meso Scale Discovery VPLEX Plus HUM IL17C
SDP
Catalog No. 5000081396
Encompass
The Meso Scale Discovery V-PLEX Human IL-17C Kit is an electrochemiluminescent immunoassay for measuring human interleukin 17C (IL-17C) in various biological fluids It uses MSD s V-PLEX technology for high sensitivity and a wide dynamic range The kit includes a 96-well plate with capture antibodies for IL-17C detection antibodies with an electrochemiluminescent label and calibrator diluents for a standard curve It detects IL-17C from 19 pg/mL to 10 000 pg/mL
Catalog No. 50-000-81396 Supplier Meso Scale Discovery Supplier No. K151WPG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-17C Kit is an electrochemiluminescent immunoassay for measuring human interleukin 17C (IL-17C) in various biological fluids It uses MSD s V-PLEX technology for high sensitivity and a wide dynamic range The kit includes a 96-well plate with capture antibodies for IL-17C detection antibodies with an electrochemiluminescent label and calibrator diluents for a standard curve It detects IL-17C from 19 pg/mL to 10 000 pg/mL