Learn More
Meso Scale Discovery VPLEX Plus HUM IL17D
SDP
Catalog No. 5000081895
Encompass
The Meso Scale Discovery V-PLEX Human IL-17D Kit is an electrochemiluminescent immunoassay for quantifying human interleukin 17D (IL-17D) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive IL-17D measurement with detection limits from 0 03 to 2500 pg/mL The kit includes a 96-well plate detection antibody calibrators and necessary reagents The V-PLEX platform offers a no-wash protocol and results in 3 5 hours enabling high-throughput quantification of this cytokine
Catalog No. 50-000-81895 Supplier Meso Scale Discovery Supplier No. K151WQG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-17D Kit is an electrochemiluminescent immunoassay for quantifying human interleukin 17D (IL-17D) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive IL-17D measurement with detection limits from 0 03 to 2500 pg/mL The kit includes a 96-well plate detection antibody calibrators and necessary reagents The V-PLEX platform offers a no-wash protocol and results in 3 5 hours enabling high-throughput quantification of this cytokine