Learn More
Meso Scale Discovery VPLEX Plus HUM IL1RA
SDP
Catalog No. 5000082400
Encompass
The V-PLEX Plus Human IL-1ra Kit from Meso Scale Discovery is a sensitive immunoassay for measuring human interleukin-1 receptor antagonist (IL-1ra) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for accurate results in 3 5 hours The assay detects as low as 19 5 pg/mL and up to 10 000 pg/mL allowing precise IL-1ra quantification The kit includes a 96-well plate with IL-1ra capture antibody detection antibody read buffer wash buffer calibrator diluent and lyophilized calibrator
Catalog No. 50-000-82400 Supplier Meso Scale Discovery Supplier No. K151WTG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human IL-1ra Kit from Meso Scale Discovery is a sensitive immunoassay for measuring human interleukin-1 receptor antagonist (IL-1ra) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for accurate results in 3 5 hours The assay detects as low as 19 5 pg/mL and up to 10 000 pg/mL allowing precise IL-1ra quantification The kit includes a 96-well plate with IL-1ra capture antibody detection antibody read buffer wash buffer calibrator diluent and lyophilized calibrator