Learn More
Meso Scale Discovery VPLEX Plus HUM IL9 Kit 25 PL
SDP
Catalog No. 5000081993
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-6 Kit is a multiplex immunoassay that quantitatively measures human interleukin 6 (IL-6) in cell culture supernatants serum plasma and other biological fluids This assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators controls assay and wash buffers and a read buffer The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81993 Supplier Meso Scale Discovery Supplier No. K151XCG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human IL-6 Kit is a multiplex immunoassay that quantitatively measures human interleukin 6 (IL-6) in cell culture supernatants serum plasma and other biological fluids This assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators controls assay and wash buffers and a read buffer The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL