Learn More
Meso Scale Discovery VPLEX Plus HUM Insulin
SDP
Catalog No. 5000078562
Encompass
The Meso Scale Discovery V-PLEX Plus Human Insulin Kit is an electrochemiluminescent immunoassay for measuring human insulin in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay range is 3 13-1000 pg/mL and takes about 3 5 hours It uses MSD s SULFO-TAG labels and MESO QuickPlex SQ 120 for detection The kit provides reagents for one 96-well plate allowing 40 samples in duplicate
Catalog No. 50-000-78562 Supplier Meso Scale Discovery Supplier No. K151S5G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human Insulin Kit is an electrochemiluminescent immunoassay for measuring human insulin in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay range is 3 13-1000 pg/mL and takes about 3 5 hours It uses MSD s SULFO-TAG labels and MESO QuickPlex SQ 120 for detection The kit provides reagents for one 96-well plate allowing 40 samples in duplicate