Learn More
Meso Scale Discovery VPLEX Plus HUM Leptin
SDP
Catalog No. 5000080342
Encompass
The Meso Scale Discovery V-PLEX Plus Human Leptin Kit is a multiplex immunoassay that quantitatively measures human leptin in serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It allows simultaneous measurement of leptin and up to three other analytes The assay takes 3 5 hours to complete and requires a MSD instrument such as SECTOR Imager QuickPlex SQ 120 or QuickPlex SQ 120
Catalog No. 50-000-80342 Supplier Meso Scale Discovery Supplier No. K151V5G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human Leptin Kit is a multiplex immunoassay that quantitatively measures human leptin in serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It allows simultaneous measurement of leptin and up to three other analytes The assay takes 3 5 hours to complete and requires a MSD instrument such as SECTOR Imager QuickPlex SQ 120 or QuickPlex SQ 120