Learn More
Meso Scale Discovery VPLEX Plus HUM PP Kit 25 PL
SDP
Catalog No. 5000080479
Encompass
The Meso Scale Discovery V-PLEX Human Pro-Inflammatory Panel 1 (4-Plex) is an electrochemiluminescent multiplex assay kit for quantifying four pro-inflammatory biomarkers IFN- IL-1 IL-6 and TNF- in human samples It uses MSD s multi-array technology for sensitive reproducible results with higher throughput than ELISA The V-PLEX assays run on MSD s SECTOR Imager reading up to 96 wells The kit includes a plate with capture antibodies detection antibodies calibrators controls diluent wash buffer read buffer and plate seals for one 96-well plate
Catalog No. 50-000-80479 Supplier Meso Scale Discovery Supplier No. K151Y5G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human Pro-Inflammatory Panel 1 (4-Plex) is an electrochemiluminescent multiplex assay kit for quantifying four pro-inflammatory biomarkers IFN- IL-1 IL-6 and TNF- in human samples It uses MSD s multi-array technology for sensitive reproducible results with higher throughput than ELISA The V-PLEX assays run on MSD s SECTOR Imager reading up to 96 wells The kit includes a plate with capture antibodies detection antibodies calibrators controls diluent wash buffer read buffer and plate seals for one 96-well plate