Learn More
Meso Scale Discovery VPLEX Plus Metabolic PNL 1 NHP
SDP
Catalog No. 5000081510
Encompass
The Meso Scale Discovery V-PLEX Non-Human Primate Glucagon Kit is an electrochemiluminescent immunoassay for the quantitative measurement of glucagon in non-human primate serum plasma and cell culture supernatants The assay utilizes MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of 78-20 000 pg/mL and shows no cross-reactivity with other analytes The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker proteins calibrator diluent and a glucagon calibrator The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-81510 Supplier Meso Scale Discovery Supplier No. K15332G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Non-Human Primate Glucagon Kit is an electrochemiluminescent immunoassay for the quantitative measurement of glucagon in non-human primate serum plasma and cell culture supernatants The assay utilizes MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of 78-20 000 pg/mL and shows no cross-reactivity with other analytes The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker proteins calibrator diluent and a glucagon calibrator The assay takes approximately 3 5 hours to complete