Learn More
Meso Scale Discovery VPLEX Plus Metabolic PNL 1 rat
SDP
Catalog No. 5000082214
Encompass
The V-PLEX Plus Metabolic Panel 1 Rat Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows simultaneous measurement of 10 rat metabolic biomarkers (albumin ALP ALT AST BUN/urea creatinine glucose total protein and uric acid) from a single small sample volume This electrochemiluminescent assay utilizes multi-array technology and provides a rapid sensitive and reproducible method to profile the health and function of tissues and organs involved in metabolism and homeostasis in rat models With a streamlined workflow requiring only 50 L of sample this kit serves as an efficient tool for assessing toxicity and efficacy in preclinical drug development
Catalog No. 50-000-82214 Supplier Meso Scale Discovery Supplier No. K15334G4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Metabolic Panel 1 Rat Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows simultaneous measurement of 10 rat metabolic biomarkers (albumin ALP ALT AST BUN/urea creatinine glucose total protein and uric acid) from a single small sample volume This electrochemiluminescent assay utilizes multi-array technology and provides a rapid sensitive and reproducible method to profile the health and function of tissues and organs involved in metabolism and homeostasis in rat models With a streamlined workflow requiring only 50 L of sample this kit serves as an efficient tool for assessing toxicity and efficacy in preclinical drug development