Learn More
Meso Scale Discovery VPLEX Plus Metabolic PNL human
SDP
Catalog No. 5000078262
Encompass
The Meso Scale Discovery V-PLEX Plus Metabolic Panel 1 Human Kit quantifies insulin leptin resistin PAI-1 active and MCP-1 in human serum plasma and cell culture supernatants This ECL-based assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from a small sample The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium read buffer calibrators and controls It can be automated with a hands-on time of 2 5 hours and requires a 50 L sample
Catalog No. 50-000-78262 Supplier Meso Scale Discovery Supplier No. K15325G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Metabolic Panel 1 Human Kit quantifies insulin leptin resistin PAI-1 active and MCP-1 in human serum plasma and cell culture supernatants This ECL-based assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from a small sample The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium read buffer calibrators and controls It can be automated with a hands-on time of 2 5 hours and requires a 50 L sample