Learn More
Meso Scale Discovery VPLEX Plus Metabolic PNL mouse
SDP
Catalog No. 5000082053
Encompass
The V-PLEX Plus Metabolic Panel 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit that measures 10 metabolic biomarkers in mouse serum plasma and tissue culture media The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals The assay uses electrochemiluminescence technology to provide sensitive and accurate results from just 25 L of sample The 10 analytes measured are albumin creatinine glucose cholesterol triglycerides HDL LDL ALT AST and total protein This kit allows efficient metabolic profiling of mice
Catalog No. 50-000-82053 Supplier Meso Scale Discovery Supplier No. K15333G2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Metabolic Panel 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit that measures 10 metabolic biomarkers in mouse serum plasma and tissue culture media The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals The assay uses electrochemiluminescence technology to provide sensitive and accurate results from just 25 L of sample The 10 analytes measured are albumin creatinine glucose cholesterol triglycerides HDL LDL ALT AST and total protein This kit allows efficient metabolic profiling of mice