Learn More
Meso Scale Discovery VPLEX Plus Metabolic PNL mouse
SDP
Catalog No. 5000082216
Encompass
The V-PLEX Plus Metabolic Panel 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous quantitative measurement of 10 metabolic biomarkers in mouse serum plasma and cell culture supernatants The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The 10 analytes measured are albumin creatinine glucose total cholesterol HDL LDL triglycerides AST ALT and LDH The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This kit allows efficient metabolic profiling of mouse models
Catalog No. 50-000-82216 Supplier Meso Scale Discovery Supplier No. K15333G1
The V-PLEX Plus Metabolic Panel 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous quantitative measurement of 10 metabolic biomarkers in mouse serum plasma and cell culture supernatants The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The 10 analytes measured are albumin creatinine glucose total cholesterol HDL LDL triglycerides AST ALT and LDH The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This kit allows efficient metabolic profiling of mouse models