Learn More
Meso Scale Discovery VPLEX Plus Metabolic PNL NHP
SDP
Catalog No. 5000079850
Encompass
The Meso Scale Discovery V-PLEX Non-Human Primate Insulin Kit is an electrochemiluminescent immunoassay for the quantitative measurement of insulin in non-human primate serum plasma and culture supernatants The assay has a detection range of 3 2-10 000 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator containing recombinant insulin The assay takes approximately 3 5 hours to complete and requires a Meso Scale Discovery instrument for analysis The kit includes reagents for one 96-well plate testing 40 samples in duplicate
Catalog No. 50-000-79850 Supplier Meso Scale Discovery Supplier No. K15332G1
The Meso Scale Discovery V-PLEX Non-Human Primate Insulin Kit is an electrochemiluminescent immunoassay for the quantitative measurement of insulin in non-human primate serum plasma and culture supernatants The assay has a detection range of 3 2-10 000 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator containing recombinant insulin The assay takes approximately 3 5 hours to complete and requires a Meso Scale Discovery instrument for analysis The kit includes reagents for one 96-well plate testing 40 samples in duplicate