Learn More
Meso Scale Discovery VPLEX Plus Metabolic PNL NHP
SDP
Catalog No. 5000082141
Encompass
The Meso Scale Discovery V-PLEX Metabolic Panel 1 is an assay kit for measuring metabolic biomarkers in non-human primate samples It allows simultaneous quantitation of 8 analytes including insulin leptin adiponectin resistin ghrelin GIP PYY and amylin The electrochemiluminescent assay is run on Meso Scale Discovery s QuickPlex SQ 120 instrument using a multiplex format to provide rapid results from small sample volumes The kit includes a plate coated with capture antibodies calibrators detection antibodies read buffer wash buffer and diluent It is suitable for serum plasma and tissue culture supernatant samples
Catalog No. 50-000-82141 Supplier Meso Scale Discovery Supplier No. K15332G2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Metabolic Panel 1 is an assay kit for measuring metabolic biomarkers in non-human primate samples It allows simultaneous quantitation of 8 analytes including insulin leptin adiponectin resistin ghrelin GIP PYY and amylin The electrochemiluminescent assay is run on Meso Scale Discovery s QuickPlex SQ 120 instrument using a multiplex format to provide rapid results from small sample volumes The kit includes a plate coated with capture antibodies calibrators detection antibodies read buffer wash buffer and diluent It is suitable for serum plasma and tissue culture supernatant samples