Learn More
Meso Scale Discovery VPLEX Plus Metabolic PNL rat
SDP
Catalog No. 5000079843
Encompass
The V-PLEX Plus Metabolic Panel 1 Rat Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of 10 metabolic biomarkers in rat serum plasma and tissue culture media The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10x diluent read buffer wash buffer and plate seals The assay has a short 2 5 hour protocol and a 4-log dynamic range Biomarkers measured include albumin total protein glucose creatinine BUN ALT AST ALP total bilirubin and cholesterol This kit allows efficient metabolic profiling of rat samples
Catalog No. 50-000-79843 Supplier Meso Scale Discovery Supplier No. K15334G2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Metabolic Panel 1 Rat Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of 10 metabolic biomarkers in rat serum plasma and tissue culture media The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10x diluent read buffer wash buffer and plate seals The assay has a short 2 5 hour protocol and a 4-log dynamic range Biomarkers measured include albumin total protein glucose creatinine BUN ALT AST ALP total bilirubin and cholesterol This kit allows efficient metabolic profiling of rat samples