Learn More
Meso Scale Discovery VPLEX Plus Metabolic PNL rat
SDP
Catalog No. 5000081883
Encompass
The V-PLEX Plus Metabolic Panel 1 Rat Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of 10 metabolic biomarkers in rat serum plasma and tissue culture media The kit includes a 96-well plate pre-coated with capture antibodies for insulin leptin amylin GLP-1 PYY ghrelin GIP C-peptide glucagon and resistin It utilizes electrochemiluminescence detection and provides high sensitivity broad dynamic range and requires low sample volume This kit allows efficient metabolic profiling and is suitable for studying diabetes obesity and metabolic disorders in rat models
Catalog No. 50-000-81883 Supplier Meso Scale Discovery Supplier No. K15334G1
The V-PLEX Plus Metabolic Panel 1 Rat Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of 10 metabolic biomarkers in rat serum plasma and tissue culture media The kit includes a 96-well plate pre-coated with capture antibodies for insulin leptin amylin GLP-1 PYY ghrelin GIP C-peptide glucagon and resistin It utilizes electrochemiluminescence detection and provides high sensitivity broad dynamic range and requires low sample volume This kit allows efficient metabolic profiling and is suitable for studying diabetes obesity and metabolic disorders in rat models