Learn More
Meso Scale Discovery VPLEX Plus MSE Cytokine 19Plex
SDP
Catalog No. 5000081346
Encompass
The V-PLEX Plus Mouse Cytokine 19-plex Kit from Meso Scale Discovery is a multiplex immunoassay that allows simultaneous measurement of 19 mouse cytokines and chemokines (IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IL-17A/F IFN- TNF- GM-CSF CXCL1 MCP-1 MIP-1 MIP-1 MIP-2 RANTES VEGF) from a single sample The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and requires as little as 25 L of sample It is suitable for measuring cytokines and chemokines in mouse serum plasma and cell culture supernatants
Catalog No. 50-000-81346 Supplier Meso Scale Discovery Supplier No. K15255G4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Mouse Cytokine 19-plex Kit from Meso Scale Discovery is a multiplex immunoassay that allows simultaneous measurement of 19 mouse cytokines and chemokines (IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IL-17A/F IFN- TNF- GM-CSF CXCL1 MCP-1 MIP-1 MIP-1 MIP-2 RANTES VEGF) from a single sample The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and requires as little as 25 L of sample It is suitable for measuring cytokines and chemokines in mouse serum plasma and cell culture supernatants