Learn More
Meso Scale Discovery VPLEX Plus MSE IL27p28/IL30
SDP
Catalog No. 5000081866
Encompass
The Meso Scale Discovery IL-27p28/IL-30 Kit is a highly sensitive electrochemiluminescent immunoassay for the quantitative measurement of IL-27p28 and IL-30 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide accurate and reproducible results with high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and assay diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81866 Supplier Meso Scale Discovery Supplier No. K152WXG2
The Meso Scale Discovery IL-27p28/IL-30 Kit is a highly sensitive electrochemiluminescent immunoassay for the quantitative measurement of IL-27p28 and IL-30 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide accurate and reproducible results with high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and assay diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL